Merck
  • Shape-tunable Pt-Ir alloy nanocatalysts with high performance in oxygen electrode reactions.
Shape-tunable Pt-Ir alloy nanocatalysts with high performance in oxygen electrode reactions.
Nanoscale (2016-12-24)
Tao Zhang, Shuai-Chen Li, Wei Zhu, Zhi-Ping Zhang, Jun Gu, Ya-Wen Zhang
ABSTRACT
For the first time, shape-tunable Pt-Ir alloy nanocatalysts including both single-crystalline (nano-octahedra (NOs), nano-truncated octahedra (NTOs), nanocubes (NCs)) and polycrystalline (nanocluster flowers (NCFs), nanowires (NWs), nano-short-chains (NSCs), and nano-octahedral stars (NOSs)) ones were synthesized with a facile one-pot solvothermal method, via precise control of the facet-selective agents (Br
Sigma-Aldrich
Iridium(III) acetylacetonate, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)