MilliporeSigma
  • Label-free detection of myocardial ischaemia in the perfused rat heart by spontaneous Raman spectroscopy.
Label-free detection of myocardial ischaemia in the perfused rat heart by spontaneous Raman spectroscopy.
Scientific reports (2017-02-12)
Suguru Ohira, Hideo Tanaka, Yoshinori Harada, Takeo Minamikawa, Yasuaki Kumamoto, Satoaki Matoba, Hitoshi Yaku, Tetsuro Takamatsu
ABSTRACT
Raman spectroscopy, which identifies intrinsic molecular constituents, has a potential for determining myocardial viability under label-free conditions. However, its suitability for evaluating myocardial ischaemia is undetermined. Focusing on cytochromes, i.e., representative molecules reflecting mitochondrial activity, we tested whether Raman spectroscopy is applicable for evaluating myocardial ischaemia especially during early ischaemic phase. We obtained spontaneous Raman spectra of the subepicardial myocardium in the Langendorff-perfused rat heart upon 532-nm excitation before and during the "stopped-flow," global ischaemia. Semi-quantitative values of the peak intensities at 750 and 1127 cm
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
4-(2-(6-(Dibutylamino)-2-naphthalenyl)ethenyl)-1-(3-sulfopropyl)pyridinium hydroxide inner salt, ≥95% (HPLC), solid