MilliporeSigma
  • Suppression of Aluminum Current Collector Dissolution by Protective Ceramic Coatings for Better High-Voltage Battery Performance.
Suppression of Aluminum Current Collector Dissolution by Protective Ceramic Coatings for Better High-Voltage Battery Performance.
Chemphyschem : a European journal of chemical physics and physical chemistry (2016-11-20)
Andreas Heckmann, Manuel Krott, Benjamin Streipert, Sven Uhlenbruck, Martin Winter, Tobias Placke
ABSTRACT
Batteries based on cathode materials that operate at high cathode potentials, such as LiNi
Sigma-Aldrich
Ethyl methyl sulfone, for energy applications, 97%