MilliporeSigma
  • Effects of miR-146a on the osteogenesis of adipose-derived mesenchymal stem cells and bone regeneration.
Effects of miR-146a on the osteogenesis of adipose-derived mesenchymal stem cells and bone regeneration.
Scientific reports (2017-02-17)
Qing Xie, Wei Wei, Jing Ruan, Yi Ding, Ai Zhuang, Xiaoping Bi, Hao Sun, Ping Gu, Zi Wang, Xianqun Fan
ABSTRACT
Increasing evidence has indicated that bone morphogenetic protein 2 (BMP2) coordinates with microRNAs (miRNAs) to form intracellular networks regulating mesenchymal stem cells (MSCs) osteogenesis. This study aimed to identify specific miRNAs in rat adipose-derived mesenchymal stem cells (ADSCs) during BMP2-induced osteogenesis, we selected the most significantly down-regulated miRNA, miR-146a, to systematically investigate its role in regulating osteogenesis and bone regeneration. Overexpressing miR-146a notably repressed ADSC osteogenesis, whereas knocking down miR-146a greatly promoted this process. Drosophila mothers against decapentaplegic protein 4 (SMAD4), an important co-activator in the BMP signaling pathway, was miR-146a's direct target and miR-146a exerted its repressive effect on SMAD4 through interacting with 3'-untranslated region (3'-UTR) of SMAD4 mRNA. Furthermore, knocking down SMAD4 attenuated the ability of miR-146a inhibitor to promote ADSC osteogenesis. Next, transduced ADSCs were incorporated with poly(sebacoyl diglyceride) (PSeD) porous scaffolds for repairing critical-sized cranial defect, the treatment of miR-146a inhibitor greatly enhanced ADSC-mediated bone regeneration with higher expression levels of SMAD4, Runt-related transcription factor 2 (Runx2) and Osterix in newly formed bone. In summary, our study showed that miR-146a negatively regulates the osteogenesis and bone regeneration from ADSCs both in vitro and in vivo.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SMAD4
Sigma-Aldrich
Triton X-100, laboratory grade