Merck
  • Ornithine decarboxylase regulates M1 macrophage activation and mucosal inflammation via histone modifications.
Ornithine decarboxylase regulates M1 macrophage activation and mucosal inflammation via histone modifications.
Proceedings of the National Academy of Sciences of the United States of America (2017-01-18)
Dana M Hardbower, Mohammad Asim, Paula B Luis, Kshipra Singh, Daniel P Barry, Chunying Yang, Meredith A Steeves, John L Cleveland, Claus Schneider, M Blanca Piazuelo, Alain P Gobert, Keith T Wilson
ABSTRACT
Macrophage activation is a critical step in host responses during bacterial infections. Ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine metabolism, has been well studied in epithelial cells and is known to have essential roles in many different cellular functions. However, its role in regulating macrophage function during bacterial infections is not well characterized. We demonstrate that macrophage-derived ODC is a critical regulator of M1 macrophage activation during both Helicobacter pylori and Citrobacter rodentium infection. Myeloid-specific Odc deletion significantly increased gastric and colonic inflammation, respectively, and enhanced M1 activation. Add-back of putrescine, the product of ODC, reversed the increased macrophage activation, indicating that ODC and putrescine are regulators of macrophage function. Odc-deficient macrophages had increased histone 3, lysine 4 (H3K4) monomethylation, and H3K9 acetylation, accompanied by decreased H3K9 di/trimethylation both in vivo and ex vivo in primary macrophages. These alterations in chromatin structure directly resulted in up-regulated gene transcription, especially M1 gene expression. Thus, ODC in macrophages tempers antimicrobial, M1 macrophage responses during bacterial infections through histone modifications and altered euchromatin formation, leading to the persistence and pathogenesis of these organisms.
Sigma-Aldrich
1,4-Diaminobutane-2,2,3,3-d4 dihydrochloride, 98 atom % D
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
BIX, ≥98% (HPLC)