Merck
  • Snail reprograms glucose metabolism by repressing phosphofructokinase PFKP allowing cancer cell survival under metabolic stress.
Snail reprograms glucose metabolism by repressing phosphofructokinase PFKP allowing cancer cell survival under metabolic stress.
Nature communications (2017-02-09)
Nam Hee Kim, Yong Hoon Cha, Jueun Lee, Seon-Hyeong Lee, Ji Hye Yang, Jun Seop Yun, Eunae Sandra Cho, Xianglan Zhang, Miso Nam, Nami Kim, Young-Su Yuk, So Young Cha, Yoonmi Lee, Joo Kyung Ryu, Sunghyouk Park, Jae-Ho Cheong, Sang Won Kang, Soo-Youl Kim, Geum-Sook Hwang, Jong In Yook, Hyun Sil Kim
ABSTRACT
Dynamic regulation of glucose flux between aerobic glycolysis and the pentose phosphate pathway (PPP) during epithelial-mesenchymal transition (EMT) is not well-understood. Here we show that Snail (SNAI1), a key transcriptional repressor of EMT, regulates glucose flux toward PPP, allowing cancer cell survival under metabolic stress. Mechanistically, Snail regulates glycolytic activity via repression of phosphofructokinase, platelet (PFKP), a major isoform of cancer-specific phosphofructokinase-1 (PFK-1), an enzyme involving the first rate-limiting step of glycolysis. The suppression of PFKP switches the glucose flux towards PPP, generating NADPH with increased metabolites of oxidative PPP. Functionally, dynamic regulation of PFKP significantly potentiates cancer cell survival under metabolic stress and increases metastatic capacities in vivo. Further, knockdown of PFKP rescues metabolic reprogramming and cell death induced by loss of Snail. Thus, the Snail-PFKP axis plays an important role in cancer cell survival via regulation of glucose flux between glycolysis and PPP.
Sigma-Aldrich
Anti-PFKM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)