MilliporeSigma
  • The tryptophan hydroxylase 1 (TPH1) gene, schizophrenia susceptibility, and suicidal behavior: a multi-centre case-control study and meta-analysis.
The tryptophan hydroxylase 1 (TPH1) gene, schizophrenia susceptibility, and suicidal behavior: a multi-centre case-control study and meta-analysis.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics (2009-06-16)
Peter Saetre, Per Lundmark, August Wang, Thomas Hansen, Henrik B Rasmussen, Srdjan Djurovic, Ingrid Melle, Ole A Andreassen, Thomas Werge, Ingrid Agartz, Håkan Hall, Lars Terenius, Erik G Jönsson
ABSTRACT
Serotonin (5-hydroxytryptamin; 5-HT) alternations has since long been suspected in the pathophysiology of schizophrenia. Tryptophan hydroxylase (tryptophan 5-monooxygenase; TPH) is the rate-limiting enzyme in the biosynthesis of 5-HT, and sequence variation in intron 6 of the TPH1 gene has been associated with schizophrenia. The minor allele (A) of this polymorphism (A218C) is also more frequent in patients who have attempted suicide and individuals who died by suicide, than in healthy control individuals. In an attempt to replicate previous findings, five single nucleotide polymorphisms (SNPs) were genotyped in 837 Scandinavian schizophrenia patients and 1,473 controls. Three SNPs spanning intron 6 and 7, including the A218C and A779C polymorphisms, were associated with schizophrenia susceptibility (P = 0.019). However there were no differences in allele frequencies of these loci between affected individuals having attempted suicide at least once and patients with no history of suicide attempts (P = 0.84). A systematic literature review and meta-analysis support the A218C polymorphism as a susceptibility locus for schizophrenia (odds ratio 1.17, 95% confidence interval 1.07-1.29). Association studies on suicide attempts are however conflicting (heterogeneity index I(2) = 0.54) and do not support the A218C/A779C polymorphisms being a susceptibility locus for suicidal behavior among individuals diagnosed with a psychiatric disorder (OR = 0.96 [0.80-1.16]). We conclude that the TPH1 A218/A779 locus increases the susceptibility of schizophrenia in Caucasian and Asian populations. In addition, the data at hand suggest that the locus contributes to the liability of psychiatric disorders characterized by elevated suicidal rates, rather than affecting suicidal behavior of individuals suffering from a psychiatric disorder.