MilliporeSigma
  • Host Engineering for High Quantum Efficiency Blue and White Fluorescent Organic Light-Emitting Diodes.
Host Engineering for High Quantum Efficiency Blue and White Fluorescent Organic Light-Emitting Diodes.
Advanced materials (Deerfield Beach, Fla.) (2015-06-17)
Wook Song, Inho Lee, Jun Yeob Lee
ABSTRACT
High quantum efficiency in blue and white fluorescence organic light-emitting diodes is achieved by developing a novel device architecture with fluorescent emitters doped in a thermally activated delayed fluorescent emitter as a host material.
Sigma-Aldrich
TBPe, sublimed
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$541.00