MilliporeSigma
  • Paroxetine alleviates T lymphocyte activation and infiltration to joints of collagen-induced arthritis.
Paroxetine alleviates T lymphocyte activation and infiltration to joints of collagen-induced arthritis.
Scientific reports (2017-03-30)
Qingtong Wang, Longsheng Wang, Li Wu, Mei Zhang, Shanshan Hu, Rui Wang, Yongsheng Han, Yujing Wu, Lingling Zhang, Xinming Wang, Wuyi Sun, Wei Wei
ABSTRACT
T cell infiltration to synovial tissue is an early pathogenic mechanism of rheumatoid arthritis (RA). In the present work, we reveal that G protein coupled receptor kinase 2 (GRK2) is abundantly expressed in T cells of collagen-induced arthritis (CIA). A GRK2 inhibitor, paroxetine protects the joints from inflammation and destruction, primarily through inhibition of both CD4
Sigma-Aldrich
Piperidine hydrochloride, 99%