Merck
  • The naturally occurring mutation Y197C does not affect the expression or signaling of the human histamine H
The naturally occurring mutation Y197C does not affect the expression or signaling of the human histamine H
Neuroscience letters (2017-01-28)
Cecilia Flores-Clemente, Juan Escamilla-Sánchez, Juan-Manuel Arias, José-Antonio Arias-Montaño
ABSTRACT
There is evidence for genetic polymorphism within the human histamine H
Sigma-Aldrich
Ciproxifan hydrochloride, ≥98% (HPLC), solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Immepip dihydrobromide, ≥97% (HPLC), powder