MilliporeSigma
  • Activation of the NMDA receptor-neuronal nitric oxide synthase pathway within the ventral bed nucleus of the stria terminalis mediates the negative affective component of pain.
Activation of the NMDA receptor-neuronal nitric oxide synthase pathway within the ventral bed nucleus of the stria terminalis mediates the negative affective component of pain.
Neuropharmacology (2017-03-13)
Satoshi Deyama, Yaya Sugano, Sakura Mori, Taiju Amano, Mitsuhiro Yoshioka, Katsuyuki Kaneda, Masabumi Minami
ABSTRACT
Pain consists of sensory and affective components. Although the neuronal mechanisms underlying the sensory component of pain have been studied extensively, those underlying its affective component are only beginning to be elucidated. Previously, we showed the pivotal role of the ventral part of the bed nucleus of the stria terminalis (vBNST) in the negative affective component of pain. Here, we examined the role of glutamate-nitric oxide (NO) signaling in the affective component of pain in rats using a conditioned place aversion (CPA) test. Intra-vBNST injection of either CNQX (an AMPA receptor antagonist) or MK-801 (an NMDA receptor antagonist) dose-dependently attenuated intraplantar formalin-induced CPA (F-CPA) without reducing nociceptive behaviors. In vivo microdialysis showed that extracellular oxidative NO metabolites (NOx) levels were significantly increased by intraplantar formalin injection. Intra-vBNST injection of NPLA (a selective neuronal NO synthase (nNOS) inhibitor), c-PTIO (a NO scavenger), or ZL006 (a postsynaptic density-95 (PSD-95)-nNOS interaction inhibitor) dose-dependently suppressed F-CPA without attenuating nociceptive behaviors. Intra-vBNST injection of NOR3 (a NO donor) produced CPA in a dose-dependent manner in the absence of noxious stimulation. Furthermore, whole-cell patch-clamp electrophysiology in the vBNST slices revealed that NOR3 induced depolarization of hyperpolarization-activated cation current (I
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
ZL006, ≥98% (HPLC)