MilliporeSigma
  • Evaluation of a QuEChERS-like extraction approach for the determination of PBDEs in mussels by immuno-assay-based screening methods.
Evaluation of a QuEChERS-like extraction approach for the determination of PBDEs in mussels by immuno-assay-based screening methods.
Talanta (2017-05-16)
Sara Romanelli, Francesca Bettazzi, Tania Martellini, Weilin L Shelver, Alessandra Cincinelli, Roberta Galarini, Ilaria Palchetti
ABSTRACT
A sample preparation method was evaluated for the determination of polybrominated diphenyl ethers (PBDEs) in mussel samples, by using colorimetric and electrochemical immunoassay-based screening methods. Herein, a rapid procedure based on QuEChERS-like extraction approach followed by solid phase purification was optimized for PBDE extraction from mussel samples. The detection limits for colorimetric and electrochemical immunoassays, calculated as BDE-47 equivalent concentration, were 0.6ngg
Sigma-Aldrich
2-Methylheptane, 98%