MilliporeSigma
  • Epigenetic dysregulation of ID4 predicts disease progression and treatment outcome in myeloid malignancies.
Epigenetic dysregulation of ID4 predicts disease progression and treatment outcome in myeloid malignancies.
Journal of cellular and molecular medicine (2017-04-30)
Jing-Dong Zhou, Ting-Juan Zhang, Xi-Xi Li, Ji-Chun Ma, Hong Guo, Xiang-Mei Wen, Wei Zhang, Lei Yang, Yang Yan, Jiang Lin, Jun Qian
ABSTRACT
Promoter hypermethylation-mediated inactivation of ID4 plays a crucial role in the development of solid tumours. This study aimed to investigate ID4 methylation and its clinical relevance in myeloid malignancies. ID4 hypermethylation was associated with higher IPSS scores, but was not an independent prognostic biomarker affecting overall survival (OS) in myelodysplastic syndrome (MDS). However, ID4 hypermethylation correlated with shorter OS and leukaemia-free survival (LFS) time and acted as an independent risk factor affecting OS in acute myeloid leukaemia (AML). Moreover, ID4 methylation was significantly decreased in the follow-up paired AML patients who achieved complete remission (CR) after induction therapy. Importantly, ID4 methylation was increased during MDS progression to AML and chronic phase (CP) progression to blast crisis (BC) in chronic myeloid leukaemia (CML). Epigenetic studies showed that ID4 methylation might be one of the mechanisms silencing ID4 expression in myeloid leukaemia. Functional studies in vitro showed that restoration of ID4 expression could inhibit cell proliferation and promote apoptosis in both K562 and HL60 cells. These findings indicate that ID4 acts as a tumour suppressor in myeloid malignancies, and ID4 methylation is a potential biomarker in predicting disease progression and treatment outcome.
Sigma-Aldrich
Sulfobromophthalein disodium salt hydrate, used to study hepatocyte transport functions