MilliporeSigma
  • 9-Phenanthrol modulates postinhibitory rebound and afterhyperpolarizing potentials in an excitatory motor neuron of the medicinal leech.
9-Phenanthrol modulates postinhibitory rebound and afterhyperpolarizing potentials in an excitatory motor neuron of the medicinal leech.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology (2017-05-13)
James D Angstadt, Joshua R Giordano, Alexander J Goncalves
ABSTRACT
Postinhibitory rebound (PIR) responses in leech dorsal excitatory motor neurons (cell DE-3) are eliminated by Ca
Product Description
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
9-Phenanthrol, technical grade
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid
Sigma-Aldrich
UCL 2077, ≥98% (HPLC), solid
Sigma-Aldrich
NNC 55-0396 hydrate, ≥98% (HPLC)