MilliporeSigma
  • Airborne Particulate Matter Induces Nonallergic Eosinophilic Sinonasal Inflammation in Mice.
Airborne Particulate Matter Induces Nonallergic Eosinophilic Sinonasal Inflammation in Mice.
American journal of respiratory cell and molecular biology (2017-03-01)
Murugappan Ramanathan, Nyall R London, Anuj Tharakan, Nitya Surya, Thomas E Sussan, Xiaoquan Rao, Sandra Y Lin, Elina Toskala, Sanjay Rajagopalan, Shyam Biswal
ABSTRACT
Exposure to airborne particulate matter (PM) has been linked to aggravation of respiratory symptoms, increased risk of cardiovascular disease, and all-cause mortality. Although the health effects of PM on the lower pulmonary airway have been extensively studied, little is known regarding the impact of chronic PM exposure on the upper sinonasal airway. We sought to test the impact of chronic airborne PM exposure on the upper respiratory system in vivo. Mice were subjected, by inhalation, to concentrated fine (2.5 μm) PM 6 h/d, 5 d/wk, for 16 weeks. Mean airborne fine PM concentration was 60.92 μm/m
Sigma-Aldrich
Human ALB / Serum albumin ELISA Kit