Merck
  • In- and ex-vivo molecular imaging of apoptosis to assess sensitivity of non-small cell lung cancer to EGFR inhibitors using probe-based confocal laser endomicroscopy.
In- and ex-vivo molecular imaging of apoptosis to assess sensitivity of non-small cell lung cancer to EGFR inhibitors using probe-based confocal laser endomicroscopy.
PloS one (2017-07-04)
Florian Guisier, Pierre Bohn, Maxime Patout, Nicolas Piton, Insaf Farah, Pierre Vera, Luc Thiberville, Mathieu Salaün
ABSTRACT
Prediction of treatment outcome of non-small cell lung cancer (NSCLC) with EGFR inhibitors on the basis of the genetic analysis of the tumor can be incorrect in case of rare or complex mutations, bypass molecular activation pathways, or pharmacodynamic variations. The aim of this study was to develop an ex vivo and in vivo real-time quantitative imaging test for EGFR inhibitors sensitivity assessment. Erlotinib resistant (A549, H460, H1975), insensitive (H1650) and hypersensitive (HCC827) cell lines were injected subcutaneously in Nude mice. Tumor xenografts from mice treated with Erlotinib were imaged ex vivo and in vivo using probe-based confocal laser endomicroscopy (pCLE) and NucView 488 Caspase 3 substrate, a fluorescent probe specific for the activated caspase 3. Assessment of apoptosis at 24h post treatment, both ex vivo in explanted tumor xenografts and in vivo, showed a significant difference between resistant cell lines (A549, H460 and H1975) and insensitive (H1650) or hypersensitive (HCC827) ones (p<0.05 for ex vivo imaging, p≤0.02 for in vivo imaging). There was also a significant difference between insensitive and hypersensitive cell lines, both ex vivo (p<0.05) and in vivo (p = 0.01). Real-time in vivo and ex vivo assessment of apoptosis using pCLE differentiates resistant from sensitive NSCLC xenografts to Erlotinib.
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-al, ≥95%, powder