Merck
  • Optimisation of a murine splenocyte mycobacterial growth inhibition assay using virulent Mycobacterium tuberculosis.
Optimisation of a murine splenocyte mycobacterial growth inhibition assay using virulent Mycobacterium tuberculosis.
Scientific reports (2017-06-08)
Christina Jensen, Line Lindebo Holm, Erik Svensson, Claus Aagaard, Morten Ruhwald
ABSTRACT
In the absence of a validated correlate of protection or robust animal models for human tuberculosis, Mycobacterial growth inhibition assays (MGIAs) aim to assess vaccines ability to inhibit mycobacterial growth in-vitro. We optimised a reproducible murine splenocyte MGIA based on in-vitro infection with virulent Mycobacterium tuberculosis (M.tb) Erdman. We identified splenocyte viability as a problem in state-of-art MGIA protocols, which can be improved by simple changes in culture conditions (viability increase from 21% to 46% at last day of culture). The growth inhibitory potential in mice immunised with either BCG, H56:CAF01 or H56:CAF01 administered side-by-side with BCG was significantly better compared to placebo in all groups (0.3 log
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%