MilliporeSigma
  • Metabolic changes in Medaka fish induced by cyanobacterial exposures in mesocosms: an integrative approach combining proteomic and metabolomic analyses.
Metabolic changes in Medaka fish induced by cyanobacterial exposures in mesocosms: an integrative approach combining proteomic and metabolomic analyses.
Scientific reports (2017-06-24)
Benoît Sotton, Alain Paris, Séverine Le Manach, Alain Blond, Gérard Lacroix, Alexis Millot, Charlotte Duval, Hélène Huet, Qin Qiao, Sophie Labrut, Giovanni Chiappetta, Joelle Vinh, Arnaud Catherine, Benjamin Marie
ABSTRACT
Cyanobacterial blooms pose serious threats to aquatic organisms and strongly impact the functioning of aquatic ecosystems. Due to their ability to produce a wide range of potentially bioactive secondary metabolites, so called cyanotoxins, cyanobacteria have been extensively studied in the past decades. Proteomic and metabolomic analyses provide a unique opportunity to evaluate the global response of hundreds of proteins and metabolites at a glance. In this study, we provide the first combined utilization of these methods targeted to identify the response of fish to bloom-forming cyanobacteria. Medaka fish (Oryzias latipes) were exposed for 96 hours either to a MC-producing or to a non-MC-producing strain of Microcystis aeruginosa and cellular, proteome and metabolome changes following exposure to cyanobacteria were characterized in the fish livers. The results suggest that a short-term exposure to cyanobacteria, producing or not MCs, induces sex-dependent molecular changes in medaka fish, without causing any cellular alterations. Globally, molecular entities involved in stress response, lipid metabolism and developmental processes exhibit the most contrasted changes following a cyanobacterial exposure. Moreover, it appears that proteomic and metabolomic analyses are useful tools to verify previous information and to additionally bring new horizons concerning molecular effects of cyanobacteria on fish.
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)