MilliporeSigma
  • Partition uptake of a brominated diphenyl ether by the edible plant root of white radish (Raphanus sativus L.).
Partition uptake of a brominated diphenyl ether by the edible plant root of white radish (Raphanus sativus L.).
Environmental pollution (Barking, Essex : 1987) (2017-02-09)
Chien-Ying Yang, Meei-Ling Chang, Siang Chen Wu, Yang-Hsin Shih
ABSTRACT
Polybrominated diphenyl ethers (PBDEs) are of a class of emerging contaminants. In this study, the accumulation of 4-bromodiphenyl ether (BDE-3) by different parts of a live white radish was investigated. Different cultural media (hydroponics, silica sand, and soil) were used to sustain the radish plant during its uptake and in-plant translocation of BDE-3. The results showed that BDE-3 can be translocated from the roots to the aboveground organs and the accumulated levels of BDE-3 in different parts of the white radish followed the order for the three types of cultivation: fibrous roots > peels > main roots > leaves. The results were analyzed by the aid of the partition-limited model for the plant uptake. The relevant partition coefficients (K
Sigma-Aldrich
4-Nitro-3-(trifluoromethyl)phenol, 99% (GC)