MilliporeSigma
Radical probing of spliceosome assembly.
Methods (San Diego, Calif.) (2017-07-04)
Charnpal S Grewal, Oliver A Kent, Andrew M MacMillan
ABSTRACT
Here we describe the synthesis and use of a directed hydroxyl radical probe, tethered to a pre-mRNA substrate, to map the structure of this substrate during the spliceosome assembly process. These studies indicate an early organization and proximation of conserved pre-mRNA sequences during spliceosome assembly. This methodology may be adapted to the synthesis of a wide variety of modified RNAs for use as probes of RNA structure and RNA-protein interaction.
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)