MilliporeSigma
  • Effects of mild ozonisation on gene expression and nuclear domains organization in vitro.
Effects of mild ozonisation on gene expression and nuclear domains organization in vitro.
Toxicology in vitro : an international journal published in association with BIBRA (2017-06-28)
C Scassellati, M Costanzo, B Cisterna, A Nodari, M Galiè, A Cattaneo, V Covi, G Tabaracci, C Bonvicini, M Malatesta
ABSTRACT
In the last two decades, the use of ozone (O
Sigma-Aldrich
5-Bromouridine, 98%