MilliporeSigma
  • High payload itraconazole-incorporated lipid nanoparticles with modulated release property for oral and parenteral administration.
High payload itraconazole-incorporated lipid nanoparticles with modulated release property for oral and parenteral administration.
The Journal of pharmacy and pharmacology (2017-04-20)
Alam Zeb, Omer S Qureshi, Hoo-Seong Kim, Myung-Sic Kim, Jong-Ho Kang, Jeong-Sook Park, Jin-Ki Kim
ABSTRACT
The aim of this study was to develop high payload itraconazole-incorporated lipid nanoparticles (HINP) with modulated release property using a binary mixture core of solid and liquid lipid for oral and parenteral administration. High payload itraconazole-incorporated lipid nanoparticles were prepared by hot high-pressure homogenization method using tristearin (TS) as a solid lipid, triolein (TO) as a liquid lipid and egg phosphatidylcholine/Tween 80/DSPE-PEG All HINP formulations showed particle size around 300 nm and polydispersity index below 0.3. The incorporation efficiencies of HINP formulations were above 80%, and more than 40 mg of ITZ was incorporated into each HINP formulation. In-vitro dissolution and release rate of ITZ from HINP increased as the amount of TO in lipid core increased. Compared with commercial formulations of ITZ, the pharmacokinetics of ITZ was improved after oral and parenteral administration of HINP formulations containing 0% or 10% of TO in lipid core. High payload itraconazole-incorporated lipid nanoparticles with a binary mixture lipid core have a great potential for the development of controlled release formulation of ITZ.
Product Description
Tristearin, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Triolein, European Pharmacopoeia (EP) Reference Standard