MilliporeSigma
  • Bcl-2 heterodimerizes in vivo with a conserved homolog, Bax, that accelerates programmed cell death.
Bcl-2 heterodimerizes in vivo with a conserved homolog, Bax, that accelerates programmed cell death.
Cell (1993-08-27)
Z N Oltvai, C L Milliman, S J Korsmeyer
ABSTRACT
Bcl-2 protein is able to repress a number of apoptotic death programs. To investigate the mechanism of Bcl-2's effect, we examined whether Bcl-2 interacted with other proteins. We identified an associated 21 kd protein partner, Bax, that has extensive amino acid homology with Bcl-2, focused within highly conserved domains I and II. Bax is encoded by six exons and demonstrates a complex pattern of alternative RNA splicing that predicts a 21 kd membrane (alpha) and two forms of cytosolic protein (beta and gamma). Bax homodimerizes and forms heterodimers with Bcl-2 in vivo. Overexpressed Bax accelerates apoptotic death induced by cytokine deprivation in an IL-3-dependent cell line. Overexpressed Bax also counters the death repressor activity of Bcl-2. These data suggest a model in which the ratio of Bcl-2 to Bax determines survival or death following an apoptotic stimulus.