Merck
  • The role of nitrite in sulfate radical-based degradation of phenolic compounds: An unexpected nitration process relevant to groundwater remediation by in-situ chemical oxidation (ISCO).
The role of nitrite in sulfate radical-based degradation of phenolic compounds: An unexpected nitration process relevant to groundwater remediation by in-situ chemical oxidation (ISCO).
Water research (2017-07-04)
Yuefei Ji, Lu Wang, Mengdi Jiang, Junhe Lu, Corinne Ferronato, Jean-Marc Chovelon
ABSTRACT
As promising in-situ chemical oxidation (ISCO) technologies, sulfate radical-based advanced oxidation processes (SR-AOPs) are applied in wastewater treatment and groundwater remediation in recent years. In this contribution, we report for the first time that, thermally activated persulfate oxidation of phenol in the presence of nitrite (NO
Sigma-Aldrich
4-Hydroxy-3-nitrobenzoic acid, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Chloro-2-nitrophenol, ≥97.0%