MilliporeSigma
Loss of
Disease models & mechanisms (2017-07-25)
Ekaterina Savchenko, Yajuvinder Singh, Henna Konttinen, Katarina Lejavova, Laura Mediavilla Santos, Alexandra Grubman, Virve Kärkkäinen, Velta Keksa-Goldsteine, Nikolay Naumenko, Pasi Tavi, Anthony R White, Tarja Malm, Jari Koistinaho, Katja M Kanninen
ABSTRACT
Neural stem/progenitor cells (NPCs) generate new neurons in the brain throughout an individual's lifetime in an intricate process called neurogenesis. Neurogenic alterations are a common feature of several adult-onset neurodegenerative diseases. The neuronal ceroid lipofuscinoses (NCLs) are the most common group of inherited neurodegenerative diseases that mainly affect children. Pathological features of the NCLs include accumulation of lysosomal storage material, neuroinflammation and neuronal degeneration, yet the exact cause of this group of diseases remains poorly understood. The function of the CLN5 protein, causative of the CLN5 disease form of NCL, is unknown. In the present study, we sought to examine neurogenesis in the neurodegenerative disorder caused by loss of
Product Description
Supelco
Serotonin, analytical standard