MilliporeSigma
  • Single-step generation of metal-plasma polymer multicore@shell nanoparticles from the gas phase.
Single-step generation of metal-plasma polymer multicore@shell nanoparticles from the gas phase.
Scientific reports (2017-08-19)
Pavel Solař, Oleksandr Polonskyi, Ansgar Olbricht, Alexander Hinz, Artem Shelemin, Ondřej Kylián, Andrei Choukourov, Franz Faupel, Hynek Biederman
ABSTRACT
Nanoparticles composed of multiple silver cores and a plasma polymer shell (multicore@shell) were prepared in a single step with a gas aggregation cluster source operating with Ar/hexamethyldisiloxane mixtures and optionally oxygen. The size distribution of the metal inclusions as well as the chemical composition and the thickness of the shells were found to be controlled by the composition of the working gas mixture. Shell matrices ranging from organosilicon plasma polymer to nearly stoichiometric SiO
Sigma-Aldrich
Hexamethyldisiloxane, puriss., ≥98.5% (GC)