MilliporeSigma
  • H-Abstraction by OH from Large Branched Alkanes: Overall Rate Measurements and Site-Specific Tertiary Rate Calculations.
H-Abstraction by OH from Large Branched Alkanes: Overall Rate Measurements and Site-Specific Tertiary Rate Calculations.
The journal of physical chemistry. A (2017-01-11)
Dapeng Liu, Fethi Khaled, Binod R Giri, Emmanuel Assaf, Christa Fittschen, Aamir Farooq
ABSTRACT
Reaction rate coefficients for the reaction of hydroxyl (OH) radicals with nine large branched alkanes (i.e., 2-methyl-3-ethyl-pentane, 2,3-dimethyl-pentane, 2,2,3-trimethylbutane, 2,2,3-trimethyl-pentane, 2,3,4-trimethyl-pentane, 3-ethyl-pentane, 2,2,3,4-tetramethyl-pentane, 2,2-dimethyl-3-ethyl-pentane, and 2,4-dimethyl-3-ethyl-pentane) are measured at high temperatures (900-1300 K) using a shock tube and narrow-line-width OH absorption diagnostic in the UV region. In addition, room-temperature measurements of six out of these nine rate coefficients are performed in a photolysis cell using high repetition laser-induced fluorescence of OH radicals. Our experimental results are combined with previous literature measurements to obtain three-parameter Arrhenius expressions valid over a wide temperature range (300-1300 K). The rate coefficients are analyzed using the next-nearest-neighbor (N-N-N) methodology to derive nine tertiary (T
Sigma-Aldrich
2,3,4-Trimethylpentane, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mg
Estimated to ship on June 15, 2025
from MILWAUKEE
$297.00
100 mg
Estimated to ship on June 15, 2025
from MILWAUKEE
$508.00
250 mg
Estimated to ship on June 15, 2025
from MILWAUKEE
$896.00
500 mg
Estimated to ship on January 28, 2025
from MILWAUKEE
$1,580.00
$1,374.60
1 g
In Stock
Details...
$2,730.00