MilliporeSigma
  • Circulating concentrations of growth-differentiation factor 15 in apparently healthy elderly individuals and patients with chronic heart failure as assessed by a new immunoradiometric sandwich assay.
Circulating concentrations of growth-differentiation factor 15 in apparently healthy elderly individuals and patients with chronic heart failure as assessed by a new immunoradiometric sandwich assay.
Clinical chemistry (2006-12-23)
Tibor Kempf, Rüdiger Horn-Wichmann, Georg Brabant, Timo Peter, Tim Allhoff, Gunnar Klein, Helmut Drexler, Nina Johnston, Lars Wallentin, Kai C Wollert
ABSTRACT
Growth-differentiation factor 15 (GDF15) is a member of the transforming growth factor beta (TGF-beta) cytokine superfamily. There has been increasing interest in using circulating GDF15 as a biomarker in patients, for example those with cardiovascular disease. We developed an IRMA that uses a polyclonal, affinity chromatography-purified goat antihuman GDF15 IgG antibody, assessed the preanalytic characteristics of GDF15, and determined circulating GDF15 concentrations in 429 apparently healthy elderly individuals and 153 patients with chronic heart failure (CHF). The assay had a detection limit of 20 ng/L, an intraassay imprecision of < or =10.6%, and an interassay imprecision of < or =12.2%. Specificity was demonstrated with size-exclusion chromatography, parallel measurements with polyclonal and monoclonal anti-GDF15 antibody, and lack of cross-reactivity with TGF-beta. The assay was not appreciably influenced by the anticoagulant matrix or unrelated biological substances. GDF15 was stable at room temperature for 48 h and resistant to 4 freeze-thaw cycles. Apparently healthy, elderly individuals presented with a median GDF15 concentration of 762 ng/L (25th-75th percentiles, 600-959 ng/L). GDF15 concentrations were associated with age and with cystatin C and C-reactive protein concentrations. CHF patients had increased GDF15 concentrations that were closely related to disease severity. The IRMA can detect GDF15 in human serum and plasma with excellent sensitivity and specificity. The reference limits and confounding variables defined for apparently healthy elderly individuals and the favorable preanalytic characteristics of GDF15 are expected to facilitate future studies of GDF15 as a biomarker in various disease settings, including CHF.
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 mL
In Stock
Details...
$225.00