MilliporeSigma
Robust, sensitive and facile method for detection of F
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-06-11)
P Madhusudhana Reddy, Shih-Rong Hsieh, Jem-Kun Chen, Chi-Jung Chang, Jing-Yuan Kang, Chih-Hsien Chen
ABSTRACT
Sensing of F
Sigma-Aldrich
Benzoyl isothiocyanate, 98%
Sigma-Aldrich
Benzoyl isocyanate, technical grade, 90%