MilliporeSigma
Surfactants assist in lipid extraction from wet Nannochloropsis sp.
Bioresource technology (2017-07-18)
Chongchong Wu, Ye Xiao, Weiguo Lin, Junying Zhu, Hector De la Hoz Siegler, Mingsheng Zong, Junfeng Rong
ABSTRACT
An efficient approach involving surfactant treatment, or the modification and utilization of surfactants that naturally occur in algae (algal-based surfactants), was developed to assist in the extraction of lipids from wet algae. Surfactants were found to be able to completely replace polar organic solvents in the extraction process. The highest yield of algal lipids extracted by hexane and algal-based surfactants was 78.8%, followed by 78.2% for hexane and oligomeric surfactant extraction, whereas the lipid yield extracted by hexane and ethanol was only 60.5%. In addition, the saponifiable lipids extracted by exploiting algal-based surfactants and hexane, or adding oligomeric surfactant and hexane, accounted for 78.6% and 75.4% of total algal lipids, respectively, which was more than 10% higher than the lipids extracted by hexane and ethanol. This work presents a method to extract lipids from algae using only nonpolar organic solvents, while obtaining high lipid yields and high selectivity to saponifiables.
Sigma-Aldrich
Trimethyloctadecylammonium chloride, ≥95.0% (calc. on dry substance, T)