MilliporeSigma
  • The first total synthesis and solution structure of a polypeptin, PE2, a cyclic lipopeptide with broad spectrum antibiotic activity.
The first total synthesis and solution structure of a polypeptin, PE2, a cyclic lipopeptide with broad spectrum antibiotic activity.
Organic & biomolecular chemistry (2017-08-16)
Simon J Mountford, Biswaranjan Mohanty, Kade D Roberts, Heidi H Yu, Martin J Scanlon, Roger L Nation, Tony Velkov, Jian Li, Philp E Thompson
ABSTRACT
The first total synthesis of a polypeptin, PE2, as well as its solution structure is reported. Synthesis in optically pure form confirms the proposed stereochemistry of the polypeptins at the 3-position on the 3-hydroxy depsipeptide moiety. We have also determined the NMR structure of PE2 in aqueous solution, showing it to form a stable ring conformation. The synthetic peptide shows anti-bacterial activity consistent with reports for naturally derived counterparts.
Sigma-Aldrich
Phenylsilane, 97%
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%