- Identification of Novel Serodiagnostic Signatures of Typhoid Fever Using a
Identification of Novel Serodiagnostic Signatures of Typhoid Fever Using a
Frontiers in microbiology (2017-10-04)
Thomas C Darton, Stephen Baker, Arlo Randall, Sabina Dongol, Abhilasha Karkey, Merryn Voysey, Michael J Carter, Claire Jones, Krista Trappl, Jozelyn Pablo, Chris Hung, Andy Teng, Adam Shandling, Tim Le, Cassidy Walker, Douglas Molina, Jason Andrews, Amit Arjyal, Buddha Basnyat, Andrew J Pollard, Christoph J Blohmke
PMID28970824
ABSTRACT
Current diagnostic tests for typhoid fever, the disease caused by
Sigma-Aldrich
Lipopolysaccharides from Salmonella typhosa, purified by gel-filtration chromatography