MilliporeSigma
  • Grb2-Mediated Recruitment of USP9X to LAT Enhances Themis Stability following Thymic Selection.
Grb2-Mediated Recruitment of USP9X to LAT Enhances Themis Stability following Thymic Selection.
Journal of immunology (Baltimore, Md. : 1950) (2017-09-08)
Anne Garreau, Gaëtan Blaize, Jérémy Argenty, Nelly Rouquié, Audrey Tourdès, Stephen A Wood, Abdelhadi Saoudi, Renaud Lesourne
ABSTRACT
Themis is a new component of the TCR signaling machinery that plays a critical role during T cell development. The positive selection of immature CD4
Sigma-Aldrich
PR-619, ≥95% (HPLC)