MilliporeSigma
On the viability of Mg extraction in MgMoN
Physical chemistry chemical physics : PCCP (2017-09-26)
R Verrelli, M E Arroyo-de-Dompablo, D Tchitchekova, A P Black, C Frontera, A Fuertes, M R Palacin
ABSTRACT
Layered MgMoN
Sigma-Aldrich
Lithium bis(oxalato)borate