MilliporeSigma
  • New and developing non-adrenoreceptor small molecule drugs for the treatment of asthma.
New and developing non-adrenoreceptor small molecule drugs for the treatment of asthma.
Expert opinion on pharmacotherapy (2017-01-19)
Neil C Thomson
ABSTRACT
Inhaled corticosteroids (ICS) alone or in combination with an inhaled long-acting beta
Sigma-Aldrich
Tiotropium bromide monohydrate, ≥98% (HPLC)