MilliporeSigma
  • Activation of unfolded protein response pathway is important for valproic acid mediated increase in immunoglobulin G productivity in recombinant Chinese hamster ovary cells.
Activation of unfolded protein response pathway is important for valproic acid mediated increase in immunoglobulin G productivity in recombinant Chinese hamster ovary cells.
Journal of bioscience and bioengineering (2017-06-12)
Kamal Prashad Segar, Vikas Chandrawanshi, Sarika Mehra
ABSTRACT
Process engineering to improve product quality and titers is gaining importance at late-stage cell culture process development. Valproic acid, a US Food and Drug Administration-approved histone deacetylase (HDAC) inhibitor, has been shown to improve cell culture performance with higher productivities and minimal effect on the product quality. However, the wider physiological impact of valproic acid on recombinant cells has not been investigated till date. In this study, we investigate the role of unfolded protein response pathway when immunoglobulin G (IgG)-secreting Chinese hamster ovary (CHO) cells are treated with valproic acid, resulting in a 3-fold increase in product titers and productivity. It is found that cells undergo an early transient endoplasmic reticulum (ER) stress on treatment with valproic acid, and subsequently adapt to perform as high producers. Induction of chaperones through enhanced XBP1 splicing activity and ATF6 activation suggests an increase in protein processing activity in these cells. We show that in addition to the enhanced recombinant mRNA expression of IgG heavy chain and light chain, the activation of unfolded protein response (UPR) pathway is critical to the increase in productivity of cells on valproic acid treatment. Further, upregulation of the UPR pathway is not through HDAC inhibition alone. To our knowledge, this is the first attempt to arrive at a phenotype-genotype mechanistic understanding of how valproic acid treatment enhances productivity in recombinant CHO cells.
Sigma-Aldrich
Valpromide, ≥97% (NMR)