MilliporeSigma
  • Effect of the amount of nickel sulphide, molybdenum disulphide and carbon nanosupport on a Tafel slope and overpotential optimization.
Effect of the amount of nickel sulphide, molybdenum disulphide and carbon nanosupport on a Tafel slope and overpotential optimization.
Nanotechnology (2017-04-07)
Maria Sarno, Eleonora Ponticorvo
ABSTRACT
A Tafel slope of 40 mV dec
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
1,2-Hexadecanediol, technical grade, 90%