MilliporeSigma
  • Wafer-scale production of vertical SnS multilayers for high-performing photoelectric devices.
Wafer-scale production of vertical SnS multilayers for high-performing photoelectric devices.
Nanoscale (2017-08-09)
Malkeshkumar Patel, Hong-Sik Kim, Joondong Kim
ABSTRACT
This study achieved wafer-scale, high quality tin monosulfide (SnS) layers. By using a solid-state reaction, the vertically aligned SnS layers spontaneously grew with sulphur reduction from the sputtered SnS
Sigma-Aldrich
D-(+)-Talose, ≥99.0% (HPLC)