Merck
  • Knockout of the Golgi stacking proteins GRASP55 and GRASP65 impairs Golgi structure and function.
Knockout of the Golgi stacking proteins GRASP55 and GRASP65 impairs Golgi structure and function.
Molecular biology of the cell (2017-08-18)
Michael E Bekier, Leibin Wang, Jie Li, Haoran Huang, Danming Tang, Xiaoyan Zhang, Yanzhuang Wang
ABSTRACT
Golgi reassembly stacking protein of 65 kDa (GRASP65) and Golgi reassembly stacking protein of 55 kDa (GRASP55) were originally identified as Golgi stacking proteins; however, subsequent GRASP knockdown experiments yielded inconsistent results with respect to the Golgi structure, indicating a limitation of RNAi-based depletion. In this study, we have applied the recently developed clustered regularly interspaced short palindromic repeats (CRISPR)/Cas9 technology to knock out GRASP55 and GRASP65, individually or in combination, in HeLa and HEK293 cells. We show that double knockout of GRASP proteins disperses the Golgi stack into single cisternae and tubulovesicular structures, accelerates protein trafficking, and impairs accurate glycosylation of proteins and lipids. These results demonstrate a critical role for GRASPs in maintaining the stacked structure of the Golgi, which is required for accurate posttranslational modifications in the Golgi. Additionally, the GRASP knockout cell lines developed in this study will be useful tools for studying the role of GRASP proteins in other important cellular processes.
Sigma-Aldrich
Cholera Toxin B subunit, FITC conjugate, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
CTB, ≥98% (HPLC)