Merck
  • Pharmacokinetics of posaconazole in koalas (Phascolarctos cinereus) after intravenous and oral administration.
Pharmacokinetics of posaconazole in koalas (Phascolarctos cinereus) after intravenous and oral administration.
Journal of veterinary pharmacology and therapeutics (2017-04-30)
S Gharibi, B Kimble, L Vogelnest, J Barnes, C K Stadler, M Govendir
ABSTRACT
The pharmacokinetic profile of posaconazole in clinically normal koalas (n = 8) was investigated. Single doses of posaconazole were administered intravenously (i.v.; 3 mg/kg; n = 2) or orally (p.o.; 6 mg/kg; n = 6) with serial plasma samples collected over 24 and 36 hr, respectively. Plasma concentrations of posaconazole were quantified by validated high-performance liquid chromatography. A noncompartmental pharmacokinetic analysis of data was performed. Following i.v. administration, estimates of the median (range) of plasma clearance (CL) and steady-state volume of distribution (V
Product Description
Supelco
Posaconazole, VETRANAL®, analytical standard