MilliporeSigma
  • Uptake of Gaseous Alkylamides by Suspended Sulfuric Acid Particles: Formation of Ammonium/Aminium Salts.
Uptake of Gaseous Alkylamides by Suspended Sulfuric Acid Particles: Formation of Ammonium/Aminium Salts.
Environmental science & technology (2017-09-15)
Hangfei Chen, Mingyi Wang, Lei Yao, Jianmin Chen, Lin Wang
ABSTRACT
Amides represent an important class of nitrogen-containing compounds in the atmosphere that can in theory interact with atmospheric acidic particles and contribute to secondary aerosol formation. In this study, uptake coefficients (γ) of six alkylamides (C
Sigma-Aldrich
N-Methylacetamide, ≥99%