MilliporeSigma
  • Electrical energy per order and current efficiency for electrochemical oxidation of p-chlorobenzoic acid with boron-doped diamond anode.
Electrical energy per order and current efficiency for electrochemical oxidation of p-chlorobenzoic acid with boron-doped diamond anode.
Chemosphere (2017-09-10)
Mariana Lanzarini-Lopes, Sergi Garcia-Segura, Kiril Hristovski, Paul Westerhoff
ABSTRACT
Electrochemical oxidation (EO) is an advanced oxidation process for water treatment to mineralize organic contaminants. While proven to degrade a range of emerging pollutants in water, less attention has been given to quantify the effect of operational variables such applied current density and pollutant concentration on efficiency and energy requirements. Particular figures of merit were mineralization current efficiency (MCE) and electrical energy per order (E
Sigma-Aldrich
4-Chlorobenzoic acid, 99%