MilliporeSigma
  • Superoxide generation from nNOS splice variants and its potential involvement in redox signal regulation.
Superoxide generation from nNOS splice variants and its potential involvement in redox signal regulation.
The Biochemical journal (2017-01-28)
Hideshi Ihara, Atsushi Kitamura, Shingo Kasamatsu, Tomoaki Ida, Yuki Kakihana, Hiroyasu Tsutsuki, Tomohiro Sawa, Yasuo Watanabe, Takaaki Akaike
ABSTRACT
We previously demonstrated different spacial expression profiles of the neuronal nitric oxide (NO) synthase (nNOS) splice variants nNOS-µ and nNOS-α in the brain; however, their exact functions are not fully understood. Here, we used electron paramagnetic resonance to compare the electron-uncoupling reactions of recombinant nNOS-µ and nNOS-α that generate reactive oxygen species (ROS), in this case superoxide. nNOS-µ generated 44% of the amount of superoxide that nNOS-α generated. We also evaluated the ROS production in HEK293 cells stably expressing nNOS-α and nNOS-µ by investigating these electron-uncoupling reactions as induced by calcium ionophore A23187. A23187 treatment induced greater ROS production in HEK293 cells expressing nNOS-α than those expressing nNOS-µ. Also, immunocytochemical analysis revealed that A23187-treated cells expressing nNOS-α produced more 8-nitroguanosine 3',5'-cyclic monophosphate, a second messenger in NO/ROS redox signaling, than did the cells expressing nNOS-µ. Molecular evolutionary analysis revealed that the ratio of nonsynonymous sites to synonymous sites for the nNOS-µ-specific region was higher than that for the complete gene, indicating that this region has fewer functional constraints than does the complete gene. These observations shed light on the physiological relevance of the nNOS-µ variant and may improve understanding of nNOS-dependent NO/ROS redox signaling and its pathophysiological consequences in neuronal systems.