MilliporeSigma
  • Notch2 signaling contributes to cell growth, invasion, and migration in salivary adenoid cystic carcinoma.
Notch2 signaling contributes to cell growth, invasion, and migration in salivary adenoid cystic carcinoma.
Molecular and cellular biochemistry (2015-10-03)
Jing Qu, Min Song, Jian Xie, Xiao-Yu Huang, Xiao-Meng Hu, Rui-Huan Gan, Yong Zhao, Li-Song Lin, Jiang Chen, Xu Lin, Da-Li Zheng, You-Guang Lu
ABSTRACT
Many studies have explored whether the Notch signaling pathway has a tumor-suppressive or an oncogenic role in various tumors; however, the role of the Notch signaling pathway in salivary adenoid cystic carcinoma (SACC) is still unknown. In this study, we attempt to define the role of Notch2 signaling in cell growth, invasion, and migration in SACC. We compared Notch2 expression in clinical SACC samples with that of normal samples by using immunohistochemical staining. Then, we down-regulated Notch2 expression to observe the effect of Notch2 on proliferation, invasion, migration, and the expression of known target genes of Notch signal pathway. According to our results, Notch2 expression was higher in SACC tissues compared with normal tissues. Knockdown of Notch2 inhibited cell proliferation, invasion, and migration in vitro and down-regulated the expression of HEY2 and CCND1. The results of this study suggest that Notch2 has an essential role in the cell growth, invasion, and migration of SACC. Notch2 may therefore be a potential target gene for the treatment of SACC by interfering with cell growth and metastasis.