Merck
  • Small molecule AT7867 proliferates PDX1-expressing pancreatic progenitor cells derived from human pluripotent stem cells.
Small molecule AT7867 proliferates PDX1-expressing pancreatic progenitor cells derived from human pluripotent stem cells.
Stem cell research (2017-08-27)
Azuma Kimura, Taro Toyoda, Yohei Nishi, Makoto Nasu, Akira Ohta, Kenji Osafune
ABSTRACT
While pancreatic islet transplantation achieves insulin independence in type 1 diabetes (T1D) patients, its widespread application is limited by donor tissue scarcity. Pancreatic progenitor cells (PPCs) give rise to all cell types in the pancreas during development. PPCs derived from human pluripotent stem cells have been shown to differentiate into functional β cells both in vitro and in vivo, and to reverse hyperglycemia, at least in mice. Therefore, PPCs have great potential to serve as an alternative cell source for cell therapy, and the identification of compounds that facilitate PPC proliferation could provide stable and large-scale pancreatic cell preparation systems in clinical settings. Here, we developed and performed cell-based screens to identify small molecules that induce the proliferation of hiPSC-derived PDX1-expressing PPCs. The screening identified AT7867, which promoted PPC proliferation approximately five-fold within six days through the maintenance of a high Ki67
Sigma-Aldrich
BIO, ≥98% (HPLC)