MilliporeSigma
  • Impact of instrument and column parameters on high-throughput liquid chromatography performance.
Impact of instrument and column parameters on high-throughput liquid chromatography performance.
Journal of chromatography. A (2017-07-25)
Martin Gilar, Thomas S McDonald, Fabrice Gritti
ABSTRACT
The speed and separation performance of high-throughput liquid chromatography (HT LC) was investigated. We evaluated the contributions of various experimental parameters to the total analysis cycle time, including column length (column void time), gradient delay, flow rate, auto-sampler (A/S) speed, and software related delays. The best case injection-to-injection cycle time of 22s was achieved using 12s gradient time and 2.1×20mm columns packed with 1.7μm C18 particles. The total 22s analytical duty cycle consisted of 2.5s column void time, 1.8s gradient delay, 12s gradient time, and approximately 5.7s for software setup delay time that served as column re-equilibration time. The achieved peak capacity for an alkylphenone sample was approximately 35, giving a peak production rate of 95 peaks per minute. We estimated the impact of LC system dispersion on peak capacity and peak production rate (peak capacity per unit of time). For HT LC scenarios (5-50mm columns and 4.8-120s long gradients) we observed that even a minor amount of system dispersion (2μL
Sigma-Aldrich
Valerophenone, 99%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)