MilliporeSigma
  • Cooperative interactions between VEGFR2 extracellular Ig-like subdomains ensure VEGFR2 dimerization.
Cooperative interactions between VEGFR2 extracellular Ig-like subdomains ensure VEGFR2 dimerization.
Biochimica et biophysica acta (2017-08-30)
Christopher King, Daniel Wirth, Samuel Workman, Kalina Hristova
ABSTRACT
Prior studies have suggested that the interactions occurring between VEGFR2 extracellular domains in the absence of ligand are complex. Here we seek novel insights into these interactions, and into the role of the different Ig-like domains (D1 through D7) in VEGFR2 dimerization. We study the dimerization of a single amino acid mutant and of three deletion mutants in the plasma membrane using two photon microscopy and fully quantified spectral imaging. We demonstrate that a set of cooperative interactions between the different Ig-like domains ensure that VEGFR2 dimerizes with a specific affinity instead of forming oligomers. The contributions of subunits D7 and D4 seem to be the most critical, as they appear essential for strong lateral interactions and for the formation of dimers, respectively. This study provides new insights into the mechanism of VEGFR2 dimerization and activation.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate and phenol red, powder, suitable for cell culture