- Involvement of Proton-Coupled Organic Cation Antiporter in Varenicline Transport at Blood-Brain Barrier of Rats and in Human Brain Capillary Endothelial Cells.
Involvement of Proton-Coupled Organic Cation Antiporter in Varenicline Transport at Blood-Brain Barrier of Rats and in Human Brain Capillary Endothelial Cells.
Journal of pharmaceutical sciences (2017-04-30)
Toshiki Kurosawa, Kei Higuchi, Takashi Okura, Kazumasa Kobayashi, Hiroyuki Kusuhara, Yoshiharu Deguchi
PMID28454746
ABSTRACT
Varenicline is a selective partial α