MilliporeSigma
  • HPMA-Copolymer Nanocarrier Targets Tumor-Associated Macrophages in Primary and Metastatic Breast Cancer.
HPMA-Copolymer Nanocarrier Targets Tumor-Associated Macrophages in Primary and Metastatic Breast Cancer.
Molecular cancer therapeutics (2017-08-24)
Melissa N Zimel, Chloe B Horowitz, Vinagolu K Rajasekhar, Alexander B Christ, Xin Wei, Jianbo Wu, Paulina M Wojnarowicz, Dong Wang, Steven R Goldring, P Edward Purdue, John H Healey
ABSTRACT
Polymeric nanocarriers such as
Sigma-Aldrich
N-(3-Aminopropyl)methacrylamide hydrochloride, contains ≤1,000 ppm MEHQ as stabilizer, 98% (HPLC)
Sigma-Aldrich
4-Aminophenylmercuric acetate, ≥90% (titration)