MilliporeSigma
  • Neuroprotective Properties of Endocannabinoids N-Arachidonoyl Dopamine and N-Docosahexaenoyl Dopamine Examined in Neuronal Precursors Derived from Human Pluripotent Stem Cells.
Neuroprotective Properties of Endocannabinoids N-Arachidonoyl Dopamine and N-Docosahexaenoyl Dopamine Examined in Neuronal Precursors Derived from Human Pluripotent Stem Cells.
Biochemistry. Biokhimiia (2017-12-11)
E V Novosadova, E L Arsenyeva, E S Manuilova, L G Khaspekov, M Yu Bobrov, V V Bezuglov, S N Illarioshkin, I A Grivennikov
ABSTRACT
Neuroprotective properties of endocannabinoids N-arachidonoyl dopamine (NADA) and N-docosahexaenoyl dopamine (DHDA) were examined in neuronal precursor cells differentiated from human induced pluripotent stem cells and subjected to oxidative stress. Both compounds exerted neuroprotective activity, which was enhanced by elevating the concentration of the endocannabinoids within the 0.1-10 µM range. However, both agents at 10 µM concentration showed a marked toxic effect resulting in death of ~30% of the cells. Finally, antagonists of cannabinoid receptors as well as the receptor of the TRPV1 endovanilloid system did not hamper the neuroprotective effects of these endocannabinoids.
Sigma-Aldrich
Arachidonoyl dopamine, ≥98% (HPLC), ethanol solution
Sigma-Aldrich
Capsazepine, ≥98% (HPLC), solid